missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl trans-4-Hydroxycyclohexanecarboxylate 98.0+%, TCI America™
Spécifications
Spécifications
| Color | Yellow |
| Boiling Point | 144°C |
| Quantity | 25 g |
| Formula Weight | 172.22 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Ethyl trans-4-Hydroxycyclohexanecarboxylate |
Sécurité et manipulation
| TSCA | No |