missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,6-Dichloronicotinate 98.0+%, TCI America™

Numéro de catalogue. E11941G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
E11941G 1 g
E11945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. E11941G Fournisseur TCI America Code fournisseur E11941G
Il en reste null

Identifiants chimiques

CAS 58584-86-4
Molecular Formula C8H7Cl2NO2
Molecular Weight (g/mol) 220.05
MDL Number MFCD09757510
InChI Key KKYBVLDQTWQETN-UHFFFAOYSA-N
Synonym 2,6-Dichloronicotinic Acid Ethyl Ester, Ethyl 2,6-Dichloropyridine-3-carboxylate, 2,6-Dichloropyridine-3-carboxylic Acid Ethyl Ester
PubChem CID 12263987
IUPAC Name ethyl 2,6-dichloropyridine-3-carboxylate
SMILES CCOC(=O)C1=C(Cl)N=C(Cl)C=C1

Spécifications

Melting Point 52°C
Color White-Yellow
Quantity 1 g
Formula Weight 220.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2,6-Dichloronicotinate