missing translation for 'onlineSavingsMsg'
Learn More

Erythromycin, Cell Culture Grade

Catalog No. p-7042366
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6227909 10 g
AAJ6227914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6227909 Supplier Thermo Scientific Chemicals Supplier No. J6227909

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Erythromycin (E-Mycin or erythrocin), CAS # 114-07-8, is a broad-spectrum antibiotic. | CAS: 114-07-8 | C37H67NO13 | 733.94 g/mol

Erythromycin, is used as a macrolide antibiotic protein synthesis inhibitor. It is widely used in cell culture applications. Depending upon the strain of bacteria, erythromycin has been used between 50 and 200 mg/L to control bacterial growth. Erythromycin resistance can be induced in bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Erythromycin is a macrolide antibiotic that works by binding the 50S ribosomal subunit and inhibiting polypeptide formation
  • Erythromycin was isolated from Saccharopolyspora erythraea

Applications

  • Erythromycin can be used to disrupt the growth of gram-positive and gram-negative bacteria
  • Erythromycin has been shown to have motor effects in the gastrointestinal system and can be used for laboratory research on migrating motor complex activity

Chemical Identifiers

CAS 114-07-8
Molecular Formula C37H67NO13
Molecular Weight (g/mol) 733.94
MDL Number MFCD00084654
InChI Key ULGZDMOVFRHVEP-RWJQBGPGSA-N
PubChem CID 12560
ChEBI CHEBI:42355
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2O)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O

Specifications

Type Erythromycin
Physical Form Crystalline Powder
Color White
Quantity 10 g
Beilstein 75279
Merck Index 14,3681
Solubility Information Soluble in water at 2mg/ml
IUPAC Name (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-{[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-14-ethyl-7,12,13-trihydroxy-4-{[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy}-3,5,7,9,11,13-hexamethyl-1-oxacyclotetradecane-2,10-dione
Formula Weight 733.93
Grade Cell Culture
Chemical Name or Material Erythromycin
Show More Show Less
EINECSNumber 204-040-1
RTECSNumber KF4375000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only