missing translation for 'onlineSavingsMsg'
Learn More

Eriochrome Cyanine TS, Ricca Chemical

$196.50 - $196.50

Specifications

Chemical Name or Material Eriochrome Cyanine
Name Note Test Solution
CAS 7732-18-5 , 7647-14-5 , 6484-52-2 , 7697-37-2 , 3564-18-9
CAS Min % 93.0,2.33,2.33,0.28,0.07
CAS Max % 97.0,2.42,2.42,0.29,0.07
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
2903-4
View Documents
Ricca Chemical Company
29034
120 mL Natural Poly Bottle
Pack for $196.50
Only null left
 
Description

Description

Specifications

Specifications

Eriochrome Cyanine
7732-18-5 , 7647-14-5 , 6484-52-2 , 7697-37-2 , 3564-18-9
97.0,2.42,2.42,0.29,0.07
Natural Poly Bottle
Liquid
Test Solution
93.0,2.33,2.33,0.28,0.07
Indicator
Red
Videos
SDS
Documents

Documents