accessibility menu, dialog, popup

UserName

ONOONH4

CAS RN 6484-52-2

IUPAC Name: ammonium nitrate
Synonyms: ammonium nitrate|nitrate d'ammonium|nitrate of ammonia|nitrato amonico|nitric acid, ammonium salt|ammonium nitricum|ammonium saltpeter|ammonium(I) nitrate (1:1)|nitric acid ammonium salt (1:1)
Molecular Weight (g/mol): 80.04
Molecular Formula: H4N2O3
InChi Key: DVARTQFDIMZBAA-UHFFFAOYSA-O
SMILES: [NH4+].[O-][N+]([O-])=O
Grade 
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results