missing translation for 'onlineSavingsMsg'
Learn More

Eriochrome Black T Indicator in 40% Ethanol/60% Triethanolamine, Reagents

Eriochrome Black T Indicator in 40% Ethanol/60% Triethanolamine, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$197.93 - $593.68

Specifications

Chemical Name or Material Eriochrome Black T Indicator in 40% Ethanol/60% Triethanolamine
CAS 102-71-6 , 64-17-5 , 67-63-0
Synonym Water hardness indicator
Percent Purity 0.8%
Grade Reagent
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01335388
View Documents Promotion Details
N/A 100 mL
Each for $197.93
Only null left
 
01335387
View Documents Promotion Details
N/A 500 mL
Each for $593.68
Only null left
 
Description

Description

Specifications

Specifications

Eriochrome Black T Indicator in 40% Ethanol/60% Triethanolamine
Water hardness indicator
Reagent
0.9982
Liquid
102-71-6 , 64-17-5 , 67-63-0
0.8%
Glass Bottle
Dark Blue
Videos
SDS
Documents

Documents