missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eriochrome Black T Indicator in 40% Ethanol/60% Triethanolamine, Reagents
Eriochrome Black T Indicator in 40% Ethanol/60% Triethanolamine, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CE015900500C
Chemical Identifiers
| 102-71-6 , 64-17-5 , 67-63-0 | |
| 0.8% | |
| Glass Bottle | |
| Dark Blue | |
| Eriochrome Black T Indicator in 40% Ethanol/60% Triethanolamine |
| Water hardness indicator | |
| Reagent | |
| 0.9982 | |
| Liquid |
Specifications
| Eriochrome Black T Indicator in 40% Ethanol/60% Triethanolamine | |
| Water hardness indicator | |
| Reagent | |
| 0.9982 | |
| 500 mL |