missing translation for 'onlineSavingsMsg'
Learn More

Eriochrome Black T Indicator in 40% Ethanol/60% Triethanolamine, Reagents

Catalog No. 01335388
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335388 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335388 Supplier Labchem Inc Supplier No. CE015900100C
Only null left

Eriochrome Black T Indicator in 40% Ethanol/60% Triethanolamine, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Eriochrome Black T Indicator in 40% Ethanol/60% Triethanolamine
CAS 102-71-6, 64-17-5, 67-63-0
Synonym Water hardness indicator
Percent Purity 0.8%
Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.9982
Color Dark Blue
Quantity 100 mL
Physical Form Liquid