missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eriochrome Black T Indicator, 1% (w/w) in Sodium Chloride, Ricca Chemical
$95.77 - $305.67
Chemical Identifiers
| CAS | 1787-61-7 , 7647-14-5 |
|---|---|
| Molecular Formula | C20H12N3NaO7S |
| Molecular Weight (g/mol) | Mixture |
| MDL Number | MFCD00003935 |
| InChI Key | JHUJLRKQZAPSDP-GXTSIBQPSA-M |
| PubChem CID | 87355429 |
| IUPAC Name | sodium (4Z)-4-[2-(1-hydroxynaphthalen-2-yl)hydrazin-1-ylidene]-7-nitro-3-oxo-3,4-dihydronaphthalene-1-sulfonate |
| SMILES | [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O |
Chemical Identifiers
| 1787-61-7 , 7647-14-5 | |
| Mixture | |
| JHUJLRKQZAPSDP-GXTSIBQPSA-M | |
| sodium (4Z)-4-[2-(1-hydroxynaphthalen-2-yl)hydrazin-1-ylidene]-7-nitro-3-oxo-3,4-dihydronaphthalene-1-sulfonate |
| C20H12N3NaO7S | |
| MFCD00003935 | |
| 87355429 | |
| [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O |
Specifications
| 1787-61-7 , 7647-14-5 | |
| C20H12N3NaO7S | |
| JHUJLRKQZAPSDP-GXTSIBQPSA-M | |
| sodium (4Z)-4-[2-(1-hydroxynaphthalen-2-yl)hydrazin-1-ylidene]-7-nitro-3-oxo-3,4-dihydronaphthalene-1-sulfonate | |
| 87355429 | |
| Indicator | |
| Gray | |
| Powder |
| 1% (w/w) | |
| MFCD00003935 | |
| [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O | |
| Mixture | |
| 1% w/w | |
| Sodium Chloride | |
| 7 | |
| Eriochrome Black T Indicator |