missing translation for 'onlineSavingsMsg'
Learn More

Eriochrome Black T Indicator, 1% (w/w) in Sodium Chloride, Ricca Chemical

Catalog No. 2900500
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
White Poly Wide Mouth Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2900500 500 g White Poly Wide Mouth Bottle
2900100 100 g White Poly Wide Mouth Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2900500 Supplier Ricca Chemical Company Supplier No. RI2900500

Chemical Identifiers

CAS 1787-61-7 , 7647-14-5
Molecular Formula C20H12N3NaO7S
Molecular Weight (g/mol) Mixture
MDL Number MFCD00003935
InChI Key JHUJLRKQZAPSDP-GXTSIBQPSA-M
PubChem CID 87355429
IUPAC Name sodium (4Z)-4-[2-(1-hydroxynaphthalen-2-yl)hydrazin-1-ylidene]-7-nitro-3-oxo-3,4-dihydronaphthalene-1-sulfonate
SMILES [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC(=CC=C12)[N+]([O-])=O)S([O-])(=O)=O

Specifications

Concentration 1% (w/w)
Percent Purity 1% w/w
Grade Indicator
Packaging White Poly Wide Mouth Bottle
Solvent or Matrix Sodium Chloride
Color Gray
pH 7
Quantity 500 g
Physical Form Powder
Chemical Name or Material Eriochrome Black T Indicator