missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6609 Hard Hat Sweatband Liner, Terry Cloth

Catalog No. 19154611
Change view
Click to view available options
Closure Type:
Snap
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type
19154611 Snap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19154611 Supplier Ergodyne™ Supplier No. 12440

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Made with absorbent terry cloth to prevent sweat from dripping into eyes when working in hot environments. Soft and low-profile, it fits comfortably under hard hats without adding bulk. This hard hat sweatband fits most hard hats and safety helmets, easily and securely attaching to the forehead of the suspension with four snap button closures.

  • Economical way for hardhat wearers to manage moisture
  • Absorbent terry cloth prevents sweat from dripping into eyes 
  • Fits most hard hats and safety helmets
  • 4 snap button closures securely attach to forehead of suspension
  • Low profile; for no-bulk wear with hard hats
  • Soft comfort; 100% cotton terry cloth 

Specifications

Product Type Hard Hat Sweatband Liner, Detachable
Material Cotton Terrycloth
Color Gray
Closure Type Snap
For Use With (Equipment) For most hard hats and safety helmets
For Use With (Application) Construction, trades, maintenance, landscaping/grounds, assembly/fabrication, material handling, freight/baggage, warehousing/distribution, Iron/Steel fabrication, for comfort and safety