missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ Evaporative Cooling Towel

Catalog No. 19154639
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19154639 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19154639 Supplier Ergodyne™ Supplier No. 12420

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dampen this advanced PVA polymer material for a towel that's significantly cooler than ambient air.

  • Advanced material absorbs and stores water without feeling sodden
  • Super-evaporative material feels significantly cooler than ambient air when wet
  • Wear around neck for hours of cooling relief or wipe face for a refreshing break

Specifications

Product Type Evaporative Cooling Towel
Color Blue
For Use With (Application) Construction, freight/baggage, drilling/mining, iron/steel work, agriculture, solar, landscaping/grounds