missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ Chill-Its™ 6700 Evaporative Cooling Tie Bandana

Catalog No. 19808109
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19808109 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19808109 Supplier Ergodyne™ Supplier No. 12305

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cool comfort for 24 to 48 hours when activated in cool water

Ideal for workers in construction, landscaping, foundries, mining, welding, factories, farming and food service.

  • Evaporative cooling works all day
  • Lightweight, comfrotable cotton construction provides all-day comfort
  • Super absorbent acrylic polymer activates easily when soaked in water for 15 mins.
  • Machine-washable; Reusable

Specifications

Material Cotton
Product Type Evaporative Cooling Bandana
Size Universal
Color Red
For Use With (Application) Ideal for workers in construction, landscaping, foundries, mining, welding, factories, farming and food service