missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erbium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$311.88 - $311.88
Chemical Identifiers
| CAS | 12061-16-4 |
|---|---|
| Molecular Formula | Er2O3 |
| Molecular Weight (g/mol) | 382.52 |
| MDL Number | MFCD00010991 |
| InChI Key | ZXGIFJXRQHZCGJ-UHFFFAOYSA-N |
| IUPAC Name | dierbium(3+) trioxidandiide |
| SMILES | [O--].[O--].[O--].[Er+3].[Er+3] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Er | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
| 12061-16-4 | |
| 382.52 | |
| ZXGIFJXRQHZCGJ-UHFFFAOYSA-N | |
| [O--].[O--].[O--].[Er+3].[Er+3] |
| Er2O3 | |
| MFCD00010991 | |
| dierbium(3+) trioxidandiide |
Specifications
| Triple-leached LDPE bottle | |
| Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
| 0.1,2.0,97.9 | |
| Er2O3 | |
| AA, ICP | |
| Certified Reference Material | |
| [O--].[O--].[O--].[Er+3].[Er+3] | |
| dierbium(3+) trioxidandiide | |
| 125 mL | |
| Matrix Concentration Cat. No. 2-5% HNO 1000μg/mL PLER2-2Y, -2X |
| 12061-16-4 , 7697-37-2 , 7732-18-5 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 0.1,2.0,97.9 | |
| MFCD00010991 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| ZXGIFJXRQHZCGJ-UHFFFAOYSA-N | |
| 2% HNO3 | |
| 382.52 | |
| Assurance | |
| Erbium Standard |
Safety and Handling
Causes skin irritation, serious eye irritation
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only