CAS RN 12061-16-4
Erbium(III) oxide, REacton™, 99.9% (REO)
CAS: 12061-16-4 Molecular Formula: Er2O3 Molecular Weight (g/mol): 382.52 MDL Number: MFCD00010991 InChI Key: ZXGIFJXRQHZCGJ-UHFFFAOYSA-N IUPAC Name: dierbium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Er+3].[Er+3]
Erbium(III) oxide, 99.8% (metals basis)
CAS: 12061-16-4 Molecular Formula: Er2O3 Molecular Weight (g/mol): 382.52 MDL Number: MFCD00010991 InChI Key: ZXGIFJXRQHZCGJ-UHFFFAOYSA-N IUPAC Name: dierbium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Er+3].[Er+3]
Erbium(III) oxide, nanopowder, 99.5% (REO)
CAS: 12061-16-4 Molecular Formula: Er2O3 Molecular Weight (g/mol): 382.52 MDL Number: MFCD00010991 InChI Key: ZXGIFJXRQHZCGJ-UHFFFAOYSA-N Synonym: Erbia IUPAC Name: dierbium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Er+3].[Er+3]
Erbium ICP Standard, 1000 ppm Er in 3% HNO3, Ricca Chemical
CAS: 12061-16-4
Erbium Standard For ICP, 1000μg/mL in 2% HNO3, SPEX CertiPrep™
CAS: 12061-16-4 Molecular Formula: Er2O3 Molecular Weight (g/mol): 382.52 MDL Number: MFCD00010991 InChI Key: ZXGIFJXRQHZCGJ-UHFFFAOYSA-N IUPAC Name: dierbium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Er+3].[Er+3]
Erbium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 12061-16-4 Molecular Formula: Er2O3 Molecular Weight (g/mol): 382.52 MDL Number: MFCD00010991 InChI Key: ZXGIFJXRQHZCGJ-UHFFFAOYSA-N IUPAC Name: dierbium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Er+3].[Er+3]