Learn More
Eppendorf™ S-4-72 Rotor Adapters
Available in a variety of configurations to suite a wide range of applications.
$681.44 - $695.25
Specifications
Type Adapter
For Use With (Equipment) S-4-72 (4 x 250mL) rotor in Model 5804/5810 centrifuges
View More Specs
Products 9
Catalog Number Mfr. No. Capacity Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Price Quantity & Availability  
5804783000
View Documents Promotion Details
Eppendorf™
5804783000
8 x 15 mL Conical Tubes
Pack of 2 Each for $681.44
Only null left
5804784006
View Documents Promotion Details
Eppendorf™
5804784006
4 x 50 mL Conical Tubes
Pack of 2 Each for $681.44
Only null left
5804787005
View Documents Promotion Details
Eppendorf™
5804787005
1 x 250 mL Tubes
Pack of 2 Each for $681.44
Only null left
5804785002
View Documents Promotion Details
Eppendorf™
5804785002
2 x 50 mL Conical Tubes
Pack of 2 Each for $685.59
Only null left
5804789008
View Documents Promotion Details
Eppendorf™
5804789008
14 x 2.6/8 mL Tubes
Pack of 2 Each for $685.59
Only null left
5804791002
View Documents Promotion Details
Eppendorf™
5804791002
13 x 5.5/12 mL Tubes
Pack of 2 Each for $685.59
Only null left
5804792009
View Documents Promotion Details
Eppendorf™
5804792009
12 x 7/17 mL Tubes
Pack of 2 Each for $685.59
Only null left
5804793005
View Documents Promotion Details
Eppendorf™
5804793005
8 x 5 mL Conical Tubes
Pack of 2 Each for $695.25
Only null left
5804794001
View Documents Promotion Details
Eppendorf™
5804794001
26 x 1.5/2 mL Tubes
Pack of 2 Each for $685.59
Only null left
Description
Description
  • For use with Centrifuge 5804/5804 R and 5810/5810 R
Specifications
Specifications
Adapter
S-4-72 (4 x 250mL) rotor in Model 5804/5810 centrifuges