Learn More
Eppendorf™ S-4-72 Rotor Adapters
Available in a variety of configurations to suite a wide range of applications.
Supplier: Eppendorf™ 5804792009
Description
- For use with Centrifuge 5804/5804 R and 5810/5810 R
Identifiants chimiques
| Adapter | |
| S-4-72 (4 x 250mL) rotor in Model 5804/5810 centrifuges |
Spécifications
| Adapter | |
| 12 x 7/17 mL Tubes | |
| S-4-72 (4 x 250mL) rotor in Model 5804/5810 centrifuges | |
| 7/17 mL |