Learn More
Eppendorf™ S-4-72 Rotor Adapters
Available in a variety of configurations to suite a wide range of applications.
Supplier:  Eppendorf™ 5804785002
 View more versions of this product
Catalog No. 5804785002
Only null left
Description
Description
  • For use with Centrifuge 5804/5804 R and 5810/5810 R
Specifications
Chemical Identifiers
Adapter
S-4-72 (4 x 250mL) rotor in Model 5804/5810 centrifuges
Specifications
Adapter
2 x 50 mL Conical Tubes
50 mL
Conical
S-4-72 (4 x 250mL) rotor in Model 5804/5810 centrifuges
Product Suggestions
Product Suggestions