Learn More
Eppendorf™ S-4-72 Rotor Adapters
Catalog No. 5804794001
Change view
Click to view available options
Capacity:
1 x 250 mL Tubes
12 x 7/17 mL Tubes
13 x 5.5/12 mL Tubes
14 x 2.6/8 mL Tubes
2 x 50 mL Conical Tubes
26 x 1.5/2 mL Tubes
4 x 50 mL Conical Tubes
8 x 15 mL Conical Tubes
8 x 5 mL Conical Tubes
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
5804794001 26 x 1.5/2 mL Tubes
5804783000 8 x 15 mL Conical Tubes
5804784006 4 x 50 mL Conical Tubes
5804787005 1 x 250 mL Tubes
5804785002 2 x 50 mL Conical Tubes
5804789008 14 x 2.6/8 mL Tubes
5804791002 13 x 5.5/12 mL Tubes
5804792009 12 x 7/17 mL Tubes
5804793005 8 x 5 mL Conical Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 5804794001 Supplier Eppendorf™ Supplier No. 5804794001
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Available in a variety of configurations to suite a wide range of applications.
  • For use with Centrifuge 5804/5804 R and 5810/5810 R
Specifications
Type Adapter
Tube Type Microtube
Capacity 26 x 1.5/2 mL Tubes
For Use With (Equipment) S-4-72 (4 x 250mL) rotor in Model 5804/5810 centrifuges
Tube Size 1.5/2 mL