Learn More
Eppendorf™ S-4-72 Rotor Adapters
Available in a variety of configurations to suite a wide range of applications.
Supplier: Eppendorf™ 5804789008
Description
- For use with Centrifuge 5804/5804 R and 5810/5810 R
Chemical Identifiers
| Adapter | |
| S-4-72 (4 x 250mL) rotor in Model 5804/5810 centrifuges |
Specifications
| Adapter | |
| 14 x 2.6/8 mL Tubes | |
| S-4-72 (4 x 250mL) rotor in Model 5804/5810 centrifuges | |
| 2.6/8 mL |