accessibility menu, dialog, popup

UserName

CAS RN 143491-57-0

IUPAC Name: 4-amino-5-fluoro-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-1,2-dihydropyrimidin-2-one
Synonyms: emtricitabine
Molecular Weight (g/mol): 247.24
Molecular Formula: C8H10FN3O3S
InChi Key: XQSPYNMVSIKCOC-NTSWFWBYSA-N
SMILES: NC1=NC(=O)N(C=C1F)[C@@H]1CS[C@H](CO)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results