missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Luminescence Enhancer For Solution Assays, Emerald-II™ Enhancer

Catalog No. T2115
Encompass
Change view
Click to view available options
Quantity:
2500 assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2115 2500 assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2115 Supplier Invitrogen™ Supplier No. T2115

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Emerald-II chemiluminescence enhancer significantly reduces aqueous quenching and tunes the wavelength of light emission.

The Emerald-II chemiluminescence enhancer significantly reduces aqueous quenching and tunes the wavelength of light emission.

  • Aqueous environments reduce chemiluminescent signal intensity of 1,2-dioxetane substrates by water-induced quenching; chemiluminescence enhancers increase the emission efficiency of light production by partitioning the water from the site of chemiluminescent signal production
  • Chemically optimized formulations enhance signal intensity in solution-based applications with dioxetane-based substrate solutions (CSPD™, CDP-Star™, Galacton™, Galacton-Plus™, Galacton-Star™, or NA-Star™ substrates)
  • Choice of Emerald-II enhancer facilitates assay development by allowing users to optimize assay signal intensity and wavelength of light emission

Optimize chemiluminescence signals

Emerald-II chemiluminescence enhancer is an essential component of tube or microplate-based assays, significantly reducing the effect of aqueous quenching. Enhancers also tune the wavelength of light emission. Emerald-II enhancer shifts the light emission maximum to 461 nm from 475 nm observed with the dioxetane alone. The use of Emerald-II enhancer can produce a wide dynamic range since photodetector saturation is less likely to occur at higher sample concentrations.

Specifications

Quantity 2500 assays
Format Liquid
Product Line EMERALD-II, NovaBright
Substrate Alkaline Phosphatase
Substrate Type AP (Alkaline Phosphatase) Substrate
Content And Storage Store the product at 4°C.
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.