missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ pRSFDuet-1 DNA Vector

Catalog No. 713413
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
713413 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 713413 Supplier MilliporeSigma™ Supplier No. 713413

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

pRSFDuet™-1 is designed for the coexpression of two target proteins. The vector encodes two multiple cloning sites (MCS) each of which is preceded by a T7 promoter, lac operator, and ribosome binding site (rbs). The vector also carries the pRSF1030 replicon (also known as NTP1), lacI gene, and kanamycin resistance marker.

Specifications

Promoter T7
Content And Storage Store at or below -70°C.
Quantity 10 μg
For Use With (Application) Coexpression of two target proteins

For Research Use Only