missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ pIEx-5 DNA Vector

Catalog No. 712423
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
712423 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 712423 Supplier MilliporeSigma™ Supplier No. 712423

missing translation for 'validMainframeMsg'

The pIEx™ vectors are designed for cloning and high-level expression of proteins in transiently transfected Spodoptera-derived insect cells. Transcription is driven by the AcNPV-derived hr5 enhancer and immediate early promoter, IE1. pIEx-5 encodes the signal peptide of adipokinetic hormone (AKH) to allow secretion of the expressed protein, and C-terminal S·Tag and His·Tag coding sequences.

Specifications

Promoter IE1
Content And Storage Store at -20°C.
Quantity 20 μg
For Use With (Application) Cloning and high-level expression of proteins in transiently transfected Spodoptera-derived insect cells

For Research Use Only