missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ pET Expression System 15b

Catalog No. 707553
Change view
Click to view available options
Quantity:
Sufficient for up to 10 transformations in each host
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
707553 Sufficient for up to 10 transformations in each host
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 707553 Supplier MilliporeSigma™ Supplier No. 707553

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The pET-15b vector carries an N-terminal His·Tag™ sequence followed by a thrombin cleavage site and three cloning sites. Note that the sequence is numbered by the pBR322 convention, so the T7 expression region is reversed on the vector map, TB045VM.

Specifications

Content And Storage Store at -70°C. Avoid freeze/thaw cycles.
Quantity Sufficient for up to 10 transformations in each host

For Research Use Only