Learn More
MilliporeSigma™ pET-46 Ek/LIC Vector Kit
Catalog No. 713353
Change view
Click to view available options
Promoter:
T7lac
Catalog No. Promoter
713353 T7lac
1 options
Catalog No. 713353 Supplier MilliporeSigma™ Supplier No. 713353
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The pET-46 Ek/LIC Vector is a LIC-compatible derivative of pET-45b(+). The plasmid contains a strong T7lac promoter and an amino-terminal His·Tag™ coding sequence immediately followed by an Ek/LIC cloning site designed to allow the generation of fusion proteins with minimal vector-encoded sequence. An optional C-terminal S·Tag™ coding sequence is compatible with purification, detection, and quantification.
Specifications
Promoter T7lac
For Research Use Only