Learn More
MilliporeSigma™ Vacuum Tubing
Catalog No. XX6700038
Change view
Click to view available options
Material:
Silicone
Catalog No. Material
XX6700038 Silicone
1 options
Catalog No. XX6700038 Supplier MilliporeSigma™ Supplier No. XX6700038
Accessories for filter holders for sample preparation.
Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel.Tri-Clamp (TC) fittings are referred to by convention as 1 1/2 in., 2 in., etc. 1 1/2 in. TC fittings connect to 1 in. or 1 1/2 in. O.D. tube sizes. 2 in. TC fittings connect to 2 in. O.D. tube size only.
Specifications
Type Laboratory Filter Holders or Cyclones
Diameter (English) Inner 0.5 in.
Overall Length (English) 10 ft.
Overall Length (Metric) 305 cm
Material Silicone