missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Heregulin-β3 Epidermal Growth Factor

Catalog No. 01201MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01201MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01201MI Supplier MilliporeSigma™ Supplier No. 01201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Heregulin-β3 protein, Epidermal growth factor (EGF) domain is available in a 100μg format.

Neuregulin 1 (NRG1) was originally identified as a 44-kD glycoprotein that interacts with the NEU/ERBB2 receptor tyrosine kinase to increase its phosphorylation on tyrosine residues. It is known that an extraordinary variety of different isoforms are produced from the NRG1 gene by alternative splicing. These isoforms include heregulins (HRGs), glial growth factors (GGFs) and sensory and motor neuron-derived factor (SMDF). They are tissue-specifically expressed and differ significantly in their structure. The HRG isoforms all contain immunoglobulin (Ig) and epidermal growth factor-like (EGF-like) domains. GGF and GGF2 isoforms contain a kringle-like sequence plus Ig and EGF-like domains; and the SMDF isoform shares only the EGF-like domain with other isoforms. The receptors for all NRG1 isoforms are the ERBB family of tyrosine kinase transmembrane receptors. Through interaction with ERBB receptors, NRG1 isoforms induce the growth and differentialtion of epithelial, neuronal, glial, and other types of cells.

Specifications

Form Lyophilized
For Use With (Application) Cell Culture
Quantity 100 μg