missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Extran™ 300 Detergent

Numéro de catalogue. MEX09961
Change view
Click to view available options
Volume (Metric):
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Volume (Metric)
MEX09961 4 L
MEX09962 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. MEX09961 Fournisseur MilliporeSigma™ Code fournisseur EX09961
Il en reste null

Powerful, liquid concentrate specially formulated to remove stubborn contamination and burnt-on residues

Can be used on equipment and apparatus made of glass, plastic, quartz, porcelain, ferrous metals and rubber.

  • Especially effective at removing organic materials such as blood and proteins
  • Works well in ultrasonic cleaning baths
  • Final rinse with distilled water will leave a residue-free surface
  • Phosphate and halogen free

Spécifications

Form Liquid
Appearance Off-White
Container Type Poly Bottle
For Use With (Application) Used on equipment and apparatus made of Glass, Plastic, Quartz, Porcelain, Ferrous Metals and Rubber
Volume (Metric) 4 L