missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Epidermal Growth Factor

Catalog No. 01407MI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01407MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01407MI Supplier MilliporeSigma™ Supplier No. 01407

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Epidermal growth factor (EGF) product is available in a 500μg format.

Epidermal growth factor has a profound effect on the differentiation of specific cells in vivo and is a potent mitogenic factor for a variety of cultured cells of both ectodermal and mesodermal origin. The EGF precursor is believed to exist as a membrane-bound molecule which is proteolytically cleaved to generate the 53-amino acid peptide hormone that stimulates cells to divide.

Specifications

Form Lyophilized
For Use With (Application) Cell Culture
Quantity 500 μg