Learn More
MilliporeSigma™ Chromolith™ Prep HPLC Columns
Catalog No. M1252520001
Change view
Click to view available options
Packing Material:
Prep RP-18 endcapped sorbent
Prep Si sorbent
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packing Material
M1252520001 Prep RP-18 endcapped sorbent
M1252510001 Prep Si sorbent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1252520001 Supplier MilliporeSigma™ Supplier No. 1.25252.0001
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Patented, monolithic silica technology allows ultra-fast and robust separations using standard HPLC systems
  • Low pressure drop at higher flow rate
  • The higher porosity assures fast adsorption and desorption kinetics
  • In comparison to particulate sorbents, monolithic column ensure shorter separation times which lead to less solvent consumption and shorter separation
  • Higher productivity and greater efficiency compared to particulate sorbents
Specifications
Diameter (Metric) 25 mm
Length (Metric) 100 mm
Pore Size 150 Å
Porosity 0.8
Quantity 1/Pk.
Surface Area 350 m2/g
Stationary Phase C18 (RP-18, ODS, Octadecyl, endcapped)
Product Line Chromolith
Packing Material Prep RP-18 endcapped sorbent
Phase Reversed Phase
Material PEEK
Technique Reversed Phase HPLC
Product Type Capillary Column
Show More Show Less