missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Casein, Bovine Milk, Protein

Catalog No. p-5364943
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21868025GM 25 g
218680100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21868025GM Supplier MilliporeSigma™ Supplier No. 21868025GM
Only null left
High molecular weight phosphoprotein. Useful as a substrate for bromelain, chymotrypsin, pepsin, trypsin, and other proteases.

Specifications

Preparation Method Dilute alkaline solutions (1mg/mL)
Quantity 25 g
Immunogen Casein
Storage Requirements Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 3 months at −20°C.
Regulatory Status RUO
Species Bovine
Recombinant Native
Form Solid