missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ BrdU Cell Proliferation Assay Kits

Catalog No. QIA58200TES
Change view
Click to view available options
Quantity:
1000 Tests
200 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QIA58200TES 200 Tests
QIA581000TE 1000 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. QIA58200TES Supplier MilliporeSigma™ Supplier No. QIA58200TEST

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BrdU Cell Proliferation Assay is a non-isotopic immunoassay for quantification of BrdU incorporation into newly synthesized DNA of actively proliferating cells. It is sensitive, rapid, and easy to perform.

BrdU Cell Proliferation Assay is a non-isotopic immunoassay for quantification of BrdU incorporation into newly synthesized DNA of actively proliferating cells. It is sensitive, rapid, and easy to perform.

Kit Components:

  • BrdU Label 58
  • Fixative/Denaturing Solution 58
  • Anti-BrdU Antibody 58
  • Antibody Diluent 58
  • TMB (Soluble)
  • ELISA Conjugate Diluent(-20C, 4%BSA)
  • ELISA Stop Solution
  • ELISA 20X Plate Wash Concentrate
  • Peroxidase Goat anti-Mouse IgG

Specifications

Format 96-well plate
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Quantity 200 Tests
For Use With (Application) Non-isotopic immunoassay for quantification of BrdU incorporation into newly synthesized DNA of actively proliferating cells

For Research Use Only