missing translation for 'onlineSavingsMsg'
Learn More

Economical pH Test Strips

Line of pH test strips incorporates the most up-to-date technology.

Supplier:  Customer Channels Group (Ccg) PH50901B50

 Afficher plus de versions de ce produit

Numéro de catalogue. S6114


Il en reste null

Description

Description

  • Nonbleed paper padded on a vinyl, 2.5 in. (6.35cm) wand
  • Stabilized to provide the most accurate color determinations
  • Color charts printed on each pack for quick and easy identification
  • Each pack includes 50 strips

Spécifications

Identifiants chimiques

Stabilized to provide user with most accurate color determination
50 Bag
pH Test Strip

Spécifications

5 to 9
1
pH Test Strip
Stabilized to provide user with most accurate color determination
50 Bag
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'shelfLife' : 3 Years

FDS
Documents

Documents

Promotions

Promotions