missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega ECL Western Blotting Substrate
Description
- Chemiluminescent Substrate to Detect Horseradish Peroxidase (HRP) Conjugates on Immunoblots
- Detect picogram levels of protein with minimal background
- Non-radioactive
- No optimization required; easily switch from other entry-level ECL substrates
Spécifications
Spécifications
| Product Type | ECL Western Blotting Substrate |
| Quantity | 500 mL |
| For Use With (Application) | Detects picogram amounts of Antigen and presence of HRP, blots can be repeteadly exposed to X-ray |
| Storage Requirements | 2°C to 10°C |