Learn More
3M™ E-A-R™ One Touch™ Pro Dispenser Refills
Description
- One gentle turn dispenses uncorded foam ear plugs one at a time
- Regular size fits most ear canals
- Shaped, self-adjusting foam provides a comfortable, low-pressure seal
- Soft foam ear plugs are easy to roll down and insert
- Refill containers are recyclable
Compliance
3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Spécifications
Spécifications
| ANSI S3.1974 | Yes |
| CSA Z94.2 | Class AL |
| Size | Universal |
| For Use With (Application) | Automotive, Construction, Food and Beverage Manufacturing, General Manufacturing, Marine, Sub Applications, Military Maintenance, Repair and Operation (MRO), Mining, Oil & Gas, Pharmaceutical, Transportation |
| High Visibility | Yes |
| Holds | 500 pairs |
| Model | 391-1012 |
| Packaging Type | Dispenser refill |
| Pairs per Package | 500 Pairs/Dispenser |
| Plug Material | Foam |
| Afficher plus de résultats |