Learn More
DuPont™ Tyvek™ IsoClean™ Lab Coat
GREENER_CHOICE
Catalog No. 17988000
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17988000 -
17988026 2X-Large
17988001 Medium
17988025 X-Large
17988024 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Should not be worn around open flame, heat source, sparks or ignition sources.
Catalog No. 17988000 Supplier DuPont™ Supplier No. IC224SWHSM00300B
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Should not be worn around open flame, heat source, sparks or ignition sources.
Protect against particulates and other contaminants in controlled environments.
  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
  • Laydown collar
  • Labcoat offers front snap closures for easy donning and doffing
  • Left chest pocket and two lower pockets
  • Raglan sleeve design for greater range of motion and comfort.
  • Bulk packaged in double transparent poly liners.
  • Manufactured under specifications that do not contain natural rubber latex
Specifications
Product Type Controlled Environments Lab Coat
Gender Unisex
Cuff Style Open
Disposable Single-use
Sterility Non-sterile
Resistance Particulates
Sleeve Style Raglan
Certifications/Compliance 1910.120 App B, 1910.1450 App A
Cleanroom Class Class 1000 (ISO 6)