Learn More
DuPont™ Tyvek™ IsoClean™ Sleeve
GREENER_CHOICE
Catalog No. 17988107
Change view
Click to view available options
Packaging:
Bulk Packaged
Clean Packed
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
17988107 Bulk Packaged
17988108 Clean Packed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 17988107 Supplier DuPont™ Supplier No. IC501BWH0001000B
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Comfortable, lightweight and durable.
Comfortable, lightweight, and durable Tyvek sleeves are available in bulk or clean packaging.
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Elastic at both ends (bicep and wrist) for arm protection
  • 18 in. long
  • One size fits most
  • Bulk packaged in double transparent poly liners
  • Manufactured under specifications that do not contain natural rubber latex
Specifications
Product Type Controlled Environment Sleeve
Size Standard
Material Tyvek
Sterility Non-sterile
Disposable Single-use
Closure Type Elastic Ends
Seam Type Bound
Cuff Style Elastic
Model IC501B
Length (English) 18 in.
Length (Metric) 45.7 cm
Packaging Bulk Packaged
Show More Show Less