Learn More
DuPont™ Tyvek™ 400 Sleeve
GREENER_CHOICE
Catalog No. 0136121
Change view
Click to view available options
Seam Type:
Serged
Catalog No. Seam Type
0136121 Serged
1 options
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 0136121 Supplier DuPont™ Supplier No. TY500SWH00020000
Edge
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Includes
Case of 100 Pair
Nonwoven Tyvek™ fabric helps resist penetration by particles and other contaminants. The inherent barrier stops submicron particles even after abrasion.
  • Elastic at both ends (bicep and wrist) for arm protection
  • 18 in. long
  • Manufactured under specifications that do not contain natural rubber latex
  • One size fits most
  • D13397569
Specifications
Product Type General Purpose Sleeve
Size Universal
Material Tyvek
Sterility Non-sterile
Disposable Single-use
Closure Type Elastic Ends
Seam Type Serged
For Use With (Application) Lead and Asbestos Abatement/Remedation, General Maintenance/Operations, Spray Painting, General Clean-Up
Length (Metric) 45.7 cm
Packaging 100 Cs.