missing translation for 'onlineSavingsMsg'
Learn More

Doxycycline Hyclate USP, Medisca™
GREENER_CHOICE

Catalog No. 18-048-041
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18048041 25 g
18048101 5 kg
18048102 100 g
18048118 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18048041 Supplier Medisca Pharmaceutique Inc Supplier No. 043404
Get 1 Extra Chemical When You Buy 4
View all promotions

missing translation for 'validMainframeMsg'

Doxycycline Hyclate USP is a yellow crystalline powder. It is hygroscopic.

Chemical Identifiers

CAS 24390-14-5

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Crystalline Powder
Color Yellow
Quantity 25 g
Percent Purity 800 to 920 μg/mg (Doxycycline)
Grade USP