missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Dog MRP2 Vesicles

Catalog No. GM0014
Encompass
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GM0014 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GM0014 Supplier Gibco™ Supplier No. GM0014

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Dog MRP2 Vesicles are insect derived purified plasma membranes with inserted MRP2 transport protein (multidrug resistance-associated protein, ABCC2).

Dog MRP2 Vesicles are insect derived purified plasma membranes with inserted MRP2 transport protein (multidrug resistance-associated protein, ABCC2). Our Dog MRP2 Vesicles can be used to evaluate test compounds and drug interactions with MRP2 in in vitro assays. MRP2 is an ATP-binding cassette (ABC) transporter that carries organic anions.

Use Dog MRP2 Vesicles to:

  • Investigate the transporter interactions of your drug candidates
  • Assess potential for transporter-mediated drug-drug interactions
  • Obtain high quality results with a large signal to noise ratios

Concept Behind ABC Transporter Vesicles
ABC transporter vesicles are easy-to-use, efficient reagents for early assessment of a drug candidate's substrate and drug interaction potential. While ABC transporters typically mediate the export of substrates out of cells, transporters expressed on these inside-out vesicles import substrates into the vesicles. It is therefore possible to quantitatively evaluate transport activity for your compound by determining the amount incorporated into the vesicles.

Clear and Reliable Results
Prepared from Sf9 cells which have been engineered to over-express specific ABC transporters, these 'inside-out' vesicles provide high levels of transporter activity with low background, giving you a clear signal if your compound is a substrate or inhibitor of a specific efflux transporter.

Transporter Vesicle Details
Dog MRP2 Vesicles are part of a larger group of transporter vesicles known as ABC transporter vesicles. They are prepared from purified plasma membranes isolated from an insect cell system (Sf9 cells transfected with baculovirus) expressing MRP2. MRP2 is expressed in various organs, and is thought to play a role in excretion of various compounds from the kidney and liver, based on its localization on the canalicular plasma membrane of hepatocytes and brush border membrane of tubular epithelial cells.

Specifications

Grade Research Grade
Quantity 0.5 mL
For Use With (Application) Pharma and Biopharma
Purity Research Grade
Packaging Vial
Content And Storage Store at -80°C.
Host Species Canine
Product Type Dog Vesicle
Format Vial
Shipping Condition Dry Ice
Physical Form Frozen
Source Insect derived purified plasma membranes with inserted MRP2 transport protein (multidrug resistance-associated protein, ABCC2)
Form Frozen
Show More Show Less
What is the supplied quantity of ATP-binding cassette (ABC) Transporter Vesicles?

The quantity supplied is 2.5 mg total protein/500 µL per vial. The protein concentration is 5 mg/mL. When our recommended protocol is followed, 50 vesicular transport assays or 125 ATPase assays can be performed with one vial of ABC Transporter Vesicles product.

What is the storage condition for ATP-binding cassette (ABC) Transporter Vesicles?

We recommend storing them at -80 degrees C.

What are ATP-binding cassette (ABC) Transporter Vesicles?

Prepared from Sf9 cells which have been engineered to over-express specific ABC transporters, ATP-binding cassette (ABC) Transporter Vesicles are “inside-out” vesicles designed for use in vesicular transport assays. They allow the investigation of possible interactions between specific ABC transporters and test compounds. ABC Transporter Vesicles provide high levels of transporter activity with low background, giving a clear signal if the test compound is a substrate or inhibitor of a specific efflux transporter.

While ABC transporters typically mediate the efflux of substrates from cells, transporters expressed on these inside-out vesicles import substrates into the vesicles. It is therefore possible to quantitatively evaluate transport activity for the test compound by determining the amount incorporated into the vesicles.
Note: One vial of vesicle product can perform 50 vesicular transport assays.

For Research Use Only. Not for use in diagnostic procedures.