missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Dipeptidyl Peptidase IV (CD26), Porcine Kidney

Catalog No. 317640250MI
Change view
Click to view available options
Quantity:
250 mIU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
317640250MI 250 mIU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 317640250MI Supplier MilliporeSigma™ Supplier No. 317640250MIU

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inhibition of DPPIV has been reported to be effective treatment for type II diabetes

  • Native, DPP IV purified from porcine kidney (renal brush border of the proximal tubule)
  • A serine exopeptidase dimer composed of two identical subunits of 110-130 kDa
  • Cleaves Xaa-Pro dipeptides from the N-terminus of oligo and polypeptides
  • DPPIV is involved in many cellular processes such as activation of cytokines, differentiation, T cell activation, and cell-matrix interactions

Specifications

Formulation In 20mM Tris-HCl, 5mM CaCl₂, 1μM ZnCl₂, 0.05% NaN₃, pH 8.0
Molecular Weight (g/mol) ∼210000-260000 (unreduced)
Purity ≥90% by SDS-PAGE
Content And Storage ≤-70°C
Form Liquid
Quantity 250 mIU
Synonym CD26, DPPIV
Product Type Dipeptidyl Peptidase IV