missing translation for 'onlineSavingsMsg'
Learn More

Diluting Fluid for Sperm, LabChem™

$55.11 - $81.21

Specifications

Chemical Name or Material Diluting Fluid for Sperm
Color Colorless
CAS Max % 94.63,5,0.37
CAS 144-55-8 , 50-00-0
Linear Formula HCHO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC135877
View Documents
LabChem
LC135877
125 mL
Each for $55.11
Only null left
 
LC135871
View Documents
LabChem
LC135871
500 mL
Each for $81.21
Only null left
 
Description

Description

Specifications

Specifications

Diluting Fluid for Sperm
94.63,5,0.37
HCHO
Soluble in water
Passes Test
1g/mL
Liquid
Colorless
144-55-8 , 50-00-0
1g/mL
30.05
Poly Bottle
Formaldehyde; Carbon monoxide; Carbon dioxide
Videos
SDS
Documents

Documents