accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Buffers and Standards
General Chemistry Solutions
Diluting Fluid for Sperm, LabChem™
LC135871
Print
Diluting Fluid for Sperm, LabChem™
Supplier:
LabChem
LC135871
View more versions of this product
Catalog No.
LC135871
Sign In
or
Register
to check your price and availability.
$81.21
/
Each
Quantity
Only null left
Explore available promotions
Add to cart
Description
Specifications
Product Suggestions
Videos
SDS
Documents
Promotions
Description
Description
Specifications
Specifications
Chemical Name or Material
Diluting Fluid for Sperm
CAS Max %
94.63,5,0.37
Linear Formula
HCHO
Solubility Information
Soluble in water
Identification
Passes Test
Specific Gravity
1g/mL
Quantity
500 mL
Color
Colorless
CAS
144-55-8
,
50-00-0
Density
1g/mL
Formula Weight
30.05
Packaging
Poly Bottle
Decomposition Information
Formaldehyde; Carbon monoxide; Carbon dioxide
Physical Form
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions