Learn More
BD Difco™ Poly Antisera
For agglutination test
Supplier:  BD 228341
 Afficher plus de versions de ce produit
Numéro de catalogue. DF2834473
Il en reste null
Description
Description
Salmonella poly antiserum
  • Febrile control antisera
  • Helps in the detection of diseases such as salmonellosis, brucellosis and rickettsial diseases
  • Qualitative and semi-quantitative slide or tube agglutination tests for homologous antibodies
  • Used along with febrile antigens as positive or negative controls
Shigella antiserum poly group
  • For identification of morphologically and biochemically identified Shigella species
  • Can be used only in slide agglutination test
  • Lyophilized, polyclonal rabbit antisera
Spécifications
Identifiants chimiques
For in vitro diagnostic use
3 mL
Spécifications
For in vitro diagnostic use
Shigella Poly Group A
3 mL
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Promotions
Promotions
For in vitro diagnostic use