Learn More
BD Difco™ Poly Antisera
For agglutination test
Specifications
Certifications/Compliance For in vitro diagnostic use
Quantity 3 mL
View More Specs
Products 4
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
DF2776473
View Documents Promotion Details
BD
227761
Shigella Poly Group A1 N/A
Only null left
DF2835472
View Documents Promotion Details
BD
228351
Shigella Poly Group B N/A
Only null left
DF2834473
View Documents Promotion Details
BD
228341
Shigella Poly Group A N/A
Only null left
DF2837470
View Documents Promotion Details
BD
228371
Shigella Poly Group D N/A
Only null left
Description
Description
Salmonella poly antiserum
  • Febrile control antisera
  • Helps in the detection of diseases such as salmonellosis, brucellosis and rickettsial diseases
  • Qualitative and semi-quantitative slide or tube agglutination tests for homologous antibodies
  • Used along with febrile antigens as positive or negative controls
Shigella antiserum poly group
  • For identification of morphologically and biochemically identified Shigella species
  • Can be used only in slide agglutination test
  • Lyophilized, polyclonal rabbit antisera
Specifications
Specifications
For in vitro diagnostic use
3 mL
Videos
SDS
Documents
Documents
For in vitro diagnostic use