missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl trans-1,2-cyclopropanedicarboxylate, 97%, Thermo Scientific™
Identifiants chimiques
| CAS | 3999-55-1 |
|---|---|
| Molecular Formula | C9H14O4 |
| Molecular Weight (g/mol) | 186.21 |
| MDL Number | MFCD00198002 |
| InChI Key | SXLDHZFJMXLFJU-RNFRBKRXSA-N |
| Synonym | diethyl trans-1,2-cyclopropanedicarboxylate, trans-diethyl cyclopropane-1,2-dicarboxylate, diethyl trans-cyclopropane-1,2-dicarboxylate, 1r,2r-diethyl cyclopropane-1,2-dicarboxylate, diethyl 1r,2r-cyclopropane-1,2-dicarboxylate, 1,2-cyclopropanedicarboxylic acid, diethyl ester, 1r,2r-rel, 1,2-cyclopropanedicarboxylicacid, 1,3-diethyl ester, 1r,2r-rel, 1s,2s-diethyl cyclopropane-1,2-dicarboxylate, diethyltrans-cyclopropane-1,2-dicarboxylate, trans-1,2-diethoxycarbonyl-cyclopropane |
| PubChem CID | 11715303 |
| IUPAC Name | diethyl (1R,2R)-cyclopropane-1,2-dicarboxylate |
| SMILES | CCOC(=O)C1CC1C(=O)OCC |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantity & Availability | ||||
|
AC169180050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
| C9H14O4 | |
| MFCD00198002 | |
| diethyl trans-1,2-cyclopropanedicarboxylate, trans-diethyl cyclopropane-1,2-dicarboxylate, diethyl trans-cyclopropane-1,2-dicarboxylate, 1r,2r-diethyl cyclopropane-1,2-dicarboxylate, diethyl 1r,2r-cyclopropane-1,2-dicarboxylate, 1,2-cyclopropanedicarboxylic acid, diethyl ester, 1r,2r-rel, 1,2-cyclopropanedicarboxylicacid, 1,3-diethyl ester, 1r,2r-rel, 1s,2s-diethyl cyclopropane-1,2-dicarboxylate, diethyltrans-cyclopropane-1,2-dicarboxylate, trans-1,2-diethoxycarbonyl-cyclopropane | |
| diethyl (1R,2R)-cyclopropane-1,2-dicarboxylate |
Spécifications
| 3999-55-1 | |
| 70°C to 75°C (1.0mmHg) | |
| Authentic | |
| C9H14O4 | |
| C3H4(CO2C2H5)2 | |
| 1.06 | |
| SXLDHZFJMXLFJU-RNFRBKRXSA-N | |
| diethyl (1R,2R)-cyclopropane-1,2-dicarboxylate | |
| 11715303 | |
| 97% |
| 1.0600g/mL | |
| 98°C | |
| 96% min. (GC) | |
| 1.4400 to 1.442 | |
| MFCD00198002 | |
| diethyl trans-1,2-cyclopropanedicarboxylate, trans-diethyl cyclopropane-1,2-dicarboxylate, diethyl trans-cyclopropane-1,2-dicarboxylate, 1r,2r-diethyl cyclopropane-1,2-dicarboxylate, diethyl 1r,2r-cyclopropane-1,2-dicarboxylate, 1,2-cyclopropanedicarboxylic acid, diethyl ester, 1r,2r-rel, 1,2-cyclopropanedicarboxylicacid, 1,3-diethyl ester, 1r,2r-rel, 1s,2s-diethyl cyclopropane-1,2-dicarboxylate, diethyltrans-cyclopropane-1,2-dicarboxylate, trans-1,2-diethoxycarbonyl-cyclopropane | |
| CCOC(=O)C1CC1C(=O)OCC | |
| 186.21 | |
| 186.21 | |
| Diethyl trans-1, 2-cyclopropanedicarboxylate |
RUO – Research Use Only