missing translation for 'onlineSavingsMsg'
Learn More

Diethyl trans-1,2-cyclopropanedicarboxylate, 97%, Thermo Scientific™

Catalog No. AC169180050
Change view
Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169180050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC169180050 Supplier Thermo Scientific Chemicals Supplier No. 169180050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 3999-55-1
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.21
MDL Number MFCD00198002
InChI Key SXLDHZFJMXLFJU-RNFRBKRXSA-N
Synonym diethyl trans-1,2-cyclopropanedicarboxylate, trans-diethyl cyclopropane-1,2-dicarboxylate, diethyl trans-cyclopropane-1,2-dicarboxylate, 1r,2r-diethyl cyclopropane-1,2-dicarboxylate, diethyl 1r,2r-cyclopropane-1,2-dicarboxylate, 1,2-cyclopropanedicarboxylic acid, diethyl ester, 1r,2r-rel, 1,2-cyclopropanedicarboxylicacid, 1,3-diethyl ester, 1r,2r-rel, 1s,2s-diethyl cyclopropane-1,2-dicarboxylate, diethyltrans-cyclopropane-1,2-dicarboxylate, trans-1,2-diethoxycarbonyl-cyclopropane
PubChem CID 11715303
IUPAC Name diethyl (1R,2R)-cyclopropane-1,2-dicarboxylate
SMILES CCOC(=O)C1CC1C(=O)OCC

Specifications

Density 1.0600g/mL
Boiling Point 70°C to 75°C (1.0mmHg)
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4400 to 1.442
Linear Formula C3H4(CO2C2H5)2
Quantity 5g
Specific Gravity 1.06
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material Diethyl trans-1, 2-cyclopropanedicarboxylate
Show More Show Less

RUO – Research Use Only