missing translation for 'onlineSavingsMsg'
Learn More

Dieldrin, Restek

Numéro de catalogue. 06719848
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
06719848 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 06719848 Fournisseur Chromatographic Code fournisseur RK 32218
Il en reste null

Identifiants chimiques

CAS 60-57-1
Molecular Formula C12H8Cl6O
Molecular Weight (g/mol) 380.895
InChI Key DFBKLUNHFCTMDC-YNXGVQRSSA-N
PubChem CID 101820673
SMILES C1C2C3C(C1C4C2O4)C5(C(=C(C3(C5(Cl)Cl)Cl)Cl)Cl)Cl

Spécifications

Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1000ug/mL, Methanol
Chemical Name or Material Dieldrin, Pesticide Mix