missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane (stabilized with 2-Methyl-2-butene) 99.5+%, TCI America™

Catalog No. D0529100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0529100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D0529100ML Supplier TCI America Supplier No. D0529100ML
Only null left

Chemical Identifiers

CAS 75-09-2
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Melting Point -97°C
Color Colorless
Boiling Point 39°C
Quantity 100 mL
UN Number 1593
Formula Weight 84.93
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material Dichloromethane (stabilized with 2-Methyl-2-butene) [for Spectrophotometry]
EINECSNumber (2)-0036
RTECSNumber PA8050000
TSCA Yes